Merck
  • A feasibility study on the multistage process for the oxalic acid pretreatment of a lignocellulosic biomass using electrodialysis.
A feasibility study on the multistage process for the oxalic acid pretreatment of a lignocellulosic biomass using electrodialysis.
Bioresource technology (2013-01-12)
Hong-Joo Lee, Sung Ju Ahn, Young-Jun Seo, Jae-Won Lee
ABSTRACT
The present study investigated the feasibility of the recovery and reuse oxalic acid in a multistage process for the pretreatment of a lignocellulosic biomass. Electrodialysis (ED), an electrochemical process using ion exchange membranes, was used to recover and reuse oxalic acid in the multistage process. The ED optimal condition for recover oxalic acid was potential of 10V and pH 2.2 in synthetic solutions. The recovery efficiency of oxalic acid from hydrolysates reached 100% at potential of 10V. The power consumption to treat 1mol of oxalic acid was estimated to be 41.0wh. At the same time, ethanol production increased up to 19g/L in the ED-treated hydrolysate, corresponding to ethanol productivity of 0.27g/L/h. It was clearly shown that bioethanol fermentation efficiency increased using the ED process, due to a small loss of fermentable sugar and a significantly high removal of inhibitory chemicals.
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, SAJ first grade, ≥97.0%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid solution, 0.5 M
Sigma-Aldrich
Oxalic acid solution, 0.05 M
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate solution, 5 mM
Sigma-Aldrich
Sodium oxalate solution, 0.0125 M
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%