Merck
  • Culture conditions and salt effects on essential oil composition of sweet marjoram (Origanum majorana) from Tunisia.
Culture conditions and salt effects on essential oil composition of sweet marjoram (Origanum majorana) from Tunisia.
Acta pharmaceutica (Zagreb, Croatia) (2012-07-04)
Olfa Baâtour, Imen Tarchoune, Hela Mahmoudi, Nawel Nassri, Wissal Abidi, Rym Kaddour, Ghaith Hamdaoui, Mouhiba Ben Nasri-Ayachi, Mohtar Lachaâl, Brahim Marzouk
ABSTRACT
O. majorana shoots were investigated for their essential oil (EO) composition. Two experiments were carried out; the first on hydroponic medium in a culture chamber and the second on inert sand in a greenhouse for 20 days. Plants were cultivated for 17 days in hydroponic medium supplemented with NaCl 100 mmol L⁻¹. The results showed that the O. majorana hydroponic medium offered higher essential oil yield than that from the greenhouse. The latter increased significantly in yield (by 50 %) under saline constraint while it did not change in the culture chamber. Under greenhouse conditions and in the absence of salt treatment, the major constituents were terpinen-4-ol and trans-sabinene hydrate. However, in the culture chamber, the major volatile components were cis-sabinene hydrate and terpinen-4-ol. In the presence of NaCl, new compounds appeared, such as eicosane, spathulenol, eugenol, and phenol. In addition, in the greenhouse, with or without salt, a very important change of trans-sabinene hydrate concentration in EO occurred, whereas in the culture chamber change appeared in cis-sabinene hydrate content.
Product Description
Terpinen 4-ol, primary reference standard
Sigma-Aldrich
4-Carvomenthenol, ≥95%, FCC, FG
Sigma-Aldrich
4-Carvomenthenol, natural, ≥95%, FG