Merck
  • An intrinsic therapy of gold nanoparticles in focal cerebral ischemia-reperfusion injury in rats.
An intrinsic therapy of gold nanoparticles in focal cerebral ischemia-reperfusion injury in rats.
Journal of biomedical nanotechnology (2013-07-19)
Zhengxia Liu, Yujie Shen, Yucheng Wu, Yujiao Yang, Jin Wu, Ping Zhou, Xiang Lu, Zhirui Guo
ABSTRACT
Ischemic stroke is a leading cause of death and disability, and the treatment options are limited. Recent studies demonstrated that the promising applications of gold nanoparticles (Au-NPs) as intrinsic therapeutics; however, little is known about the effect of Au-NPs on cerebral ischemia-reperfusion (I/R) injury. In this paper, the role of Au-NPs on cerebral I/R injury in the rat middle cerebral artery occlusion (MCAO) model was investigated using citrate-capped 5 nm and 20 nm Au-NPs. It was found that 20 nm Au-NPs administration led to remarkable amelioration of neurologic deficits and infarction volumes. Further research revealed that 20 nm Au-NPs elevated the production of anti-inflammatory cytokines, as well as inhibited I/R induced activation of astrocytes and microglias. Moreover, the expressions of anti-apoptotic proteins were up-regulated and the pro-apoptotic molecules were down-regulated in the post-ischemic brains. These effects were opposite in 5 nm group. The above results showed that the neuroprotection of 20 nm Au-NPs was attributable to its anti-inflammatory and anti-apoptotic effect. This study could provide a novel strategy for the treatment of cerebral I/R injury.
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, 99.999% (Au), layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass slide, 99.999% (Au), layer thickness 100 Å