Merck
  • Comparison of biochemical stress indicators in juvenile captive estuarine crocodiles (Crocodylus porosus) following physical restraint or chemical restraint by midazolam injection.
Comparison of biochemical stress indicators in juvenile captive estuarine crocodiles (Crocodylus porosus) following physical restraint or chemical restraint by midazolam injection.
Journal of wildlife diseases (2013-06-20)
Annabelle Olsson, David Phalen
ABSTRACT
Using a prospective, randomized study design we demonstrate that midazolam sedation minimizes acidosis compared with physical restraint in captive juvenile estuarine crocodiles during handling or noninvasive procedures at preferred body temperature. A dose of midazolam (5.0 mg/kg) was administered intramuscularly into the forelimb of 20 male estuarine crocodiles weighing 2-3.5 kg. Their heart and respiratory rate and degree of sedation were monitored until recovery and then daily for 7 subsequent days. Blood samples were taken at 30, 60, 90, 180, and 360 min. We recorded lactate, partial pressure of carbon dioxide (CO2), hematocrit, glucose, and blood pH. A second group (1.9-2.6 kg) was physically restrained for 5 min and the same parameters recorded. Physically restrained animals demonstrated elevated heart rate, respiratory rate, glucose, lactate, and anion gap compared with the midazolam-treated group. Physically restrained animals had lower pH, bicarbonate, and partial pressure of CO2 compared with the midazolam-treated group. Behavior in the physically restrained group in the days following the study was disrupted, with reluctance to feed and bask, compared with midazolam-treated animals whose behavior was normal. We conclude that midazolam administered in the forelimb of captive estuarine crocodiles of 2-3.5 kg provides predictable onset and duration of sedation enabling physical examination, sample collection, and translocation of the animals with minimal disturbance to lactate, pH, and CO2. Behavior following recovery appears normal.
Product Description
Supelco
Lactic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
DL-Lactic acid, ~90% (T)
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
DL-Lactic acid, JIS special grade, 85.0-92.0%