Merck
  • Oral chemoprevention of skin cancer in mice by benzophenone sunscreens dioxybenzone and octabenzone in drinking water.
Oral chemoprevention of skin cancer in mice by benzophenone sunscreens dioxybenzone and octabenzone in drinking water.
Anticancer research (2013-06-12)
G Subba Rao, Harukuni Tokuda, Eiichiro Ichiishi, Midori Takasaki, Akira Iida, Nobutaka Suzuki, Takao Konoshima, Govind J Kapadia
ABSTRACT
Sunscreen compounds with added benefit of skin cancer prevention have both public and commercial interests. Our earlier study using the Epstein-Barr virus early antigen in vitro assay reported on skin cancer chemoprevention potential of benzophenone sunscreens. We now report the in vivo antitumor activity of two of the benzophenone sunscreens which tested positively in the in vitro assay, octabenzone (UV-1) and dioxybenzone (UV-2), in the two-stage mouse skin carcinogenesis model using (±)-(E)-4-methyl-2-[-(E)-hydroxyamino]-5-nitro-6-methoxy-3-hexanamide (NOR-1) as inducer and 12-O-tetradecanoyl-phorbol-13-acetate (TPA) as promoter. Pathogen-free, female hairless mice of HOS:HR-1 strain, 15 animals per control and test groups, were used. Skin tumors were induced by a single dose of NOR-1 (390 nmol in 100 μl of acetone). One week later, TPA (1.7 nmol in 100 μl of acetone) was applied to skin twice weekly for 20 weeks as tumor a promoter. The test compounds UV-I or UV-2 were administered at 0.0025% to mice through drinking water ad libitum, starting one week prior to and stopping one week after tumor initiation. All animals were examined weekly for the development of skin papillomas. In both UV-1- and UV-2-treated mice, a two-week delay in tumor appearance, and significant inhibition (p<0.001) of tumor incidence (50% and 60%, respectively) and tumor burden (papilloma inhibition/mouse, 50% and 70%, respectively) were observed when compared to the positive control group. UV-2 (dihydroxy derivative) was a more potent inhibitor of skin tumor than UV-1 (monohydroxy derivative), which followed their antioxidant activity ranking. The results affirm the skin cancer chemoprevention potential of orally-ingested benzophenone sunscreens in mice and warrant studies in humans to validate synergistic protection achievable by complementation of oral and topical sunscreen usage.
Product Description
Supelco
Melting point standard 47-49°C, analytical standard
Sigma-Aldrich
2,2′-Dihydroxy-4-methoxybenzophenone, 98%
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Supelco
Dioxybenzone, analytical standard
Benzophenone, European Pharmacopoeia (EP) Reference Standard