Merck
What does S-palmitoylation do to membrane proteins?
The FEBS journal (2013-04-05)
Sanja Blaskovic, Mathieu Blanc, F Gisou van der Goot
ABSTRACT
S-palmitoylation is post-translational modification, which consists in the addition of a C16 acyl chain to cytosolic cysteines and which is unique amongst lipid modifications in that it is reversible. It can thus, like phosphorylation or ubiquitination, act as a switch. While palmitoylation of soluble proteins allows them to interact with membranes, the consequences of palmitoylation for transmembrane proteins are more enigmatic. We briefly review the current knowledge regarding the enzymes responsible for palmitate addition and removal. We then describe various observed consequences of membrane protein palmitoylation. We propose that the direct effects of palmitoylation on transmembrane proteins, however, might be limited to four non-mutually exclusive mechanistic consequences: alterations in the conformation of transmembrane domains, association with specific membrane domains, controlled interactions with other proteins and controlled interplay with other post-translational modifications.
Product Description
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, analytical standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG