Merck
  • Lithium prevents early cytosolic calcium increase and secondary injurious calcium overload in glycolytically inhibited endothelial cells.
Lithium prevents early cytosolic calcium increase and secondary injurious calcium overload in glycolytically inhibited endothelial cells.
Biochemical and biophysical research communications (2013-04-02)
Bert Bosche, Matthias Schäfer, Rudolf Graf, Frauke V Härtel, Ute Schäfer, Thomas Noll
ABSTRACT
Cytosolic free calcium concentration ([Ca(2+)]i) is a central signalling element for the maintenance of endothelial barrier function. Under physiological conditions, it is controlled within narrow limits. Metabolic inhibition during ischemia/reperfusion, however, induces [Ca(2+)]i overload, which results in barrier failure. In a model of cultured porcine aortic endothelial monolayers (EC), we addressed the question of whether [Ca(2+)]i overload can be prevented by lithium treatment. [Ca(2+)]i and ATP were analysed using Fura-2 and HPLC, respectively. The combined inhibition of glycolytic and mitochondrial ATP synthesis by 2-desoxy-d-glucose (5mM; 2-DG) plus sodium cyanide (5mM; NaCN) caused a significant decrease in cellular ATP content (14±1 nmol/mg protein vs. 18±1 nmol/mg protein in the control, n=6 culture dishes, P<0.05), an increase in [Ca(2+)]i (278±24 nM vs. 71±2 nM in the control, n=60 cells, P<0.05), and the formation of gaps between adjacent EC. These observations indicate that there is impaired barrier function at an early state of metabolic inhibition. Glycolytic inhibition alone by 10mM 2-DG led to a similar decrease in ATP content (14±2 nmol/mg vs. 18±1 nmol/mg in the control, P<0.05) with a delay of 5 min. The [Ca(2+)]i response of EC was biphasic with a peak after 1 min (183±6 nM vs. 71±1 nM, n=60 cells, P<0.05) followed by a sustained increase in [Ca(2+)]i. A 24-h pre-treatment with 10mM of lithium chloride before the inhibition of ATP synthesis abolished both phases of the 2-DG-induced [Ca(2+)]i increase. This effect was not observed when lithium chloride was added simultaneously with 2-DG. We conclude that lithium chloride abolishes the injurious [Ca(2+)]i overload in EC and that this most likely occurs by preventing inositol 3-phosphate-sensitive Ca(2+)-release from the endoplasmic reticulum. Though further research is needed, these findings provide a novel option for therapeutic strategies to protect the endothelium against imminent barrier failure.
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
2-Deoxy-D-glucose, ≥99% (GC), crystalline
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), BioXtra
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium standard solution, suitable for atomic absorption spectrometry, 1000 ppm Li, 1 mg/mL Li