Merck
  • The effects of host defence elicitors on betacyanin accumulation in Amaranthus mangostanus seedlings.
The effects of host defence elicitors on betacyanin accumulation in Amaranthus mangostanus seedlings.
Food chemistry (2013-02-28)
Shifeng Cao, Ting Liu, Yueming Jiang, Shenggen He, Dion K Harrison, Daryl C Joyce
ABSTRACT
The effect of elicitors associated with host defence on betacyanin accumulation in Amaranthus mangostanus seedlings was investigated. Under the conditions of the experiments, betacyanin accumulation was generally enhanced by light. Methyl jasmonate (MeJA) treatment increased betacyanin synthesis in a concentration-dependent response. Seedlings treated with ethylene as 5mM Ethephon also had elevated levels of betacyanin. In contrast, salicylic acid (SA) and H(2)O(2) treatments had no influence on betacyanin contents in light or dark. Combined MeJA with Ethephon or H(2)O(2) had an additive effect on betacyanin accumulation in dark-grown seedlings. However, a decline was recorded in light-grown seedlings. Moreover, an antagonistic effect on betacyanin synthesis was found when MeJA and SA were added simultaneously. Our results indicate that betacyanin content in A. mangostanus seedlings can be upregulated by MeJA and ethylene. Both additive and antagonistic effects in regulating betacyanin synthesis in A. mangostanus seedlings were observed between MeJA and other elicitors.
Product Description
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture