Merck
  • Switchable catalytic acrylamide hydrogels cross-linked by hemin/G-quadruplexes.
Switchable catalytic acrylamide hydrogels cross-linked by hemin/G-quadruplexes.
Nano letters (2013-02-21)
Chun-Hua Lu, Xiu-Juan Qi, Ron Orbach, Huang-Hao Yang, Iris Mironi-Harpaz, Dror Seliktar, Itamar Willner
ABSTRACT
Copolymer chains consisting of acrylamide units and guanine (G)-containing oligonucleotide-tethered acrylamide units undergo, in the presence of K(+) ions, cross-linking by G-quadruplexes to yield a hydrogel. The hydrogel is dissociated upon addition of 18-crown-6 ether that traps the K(+) ions. Reversible formation and dissociation of the hydrogel is demonstrated by the cyclic addition of K(+) ions and 18-crown-6 ether, respectively. Formation of the hydrogel in the presence of hemin results in a hemin/G-quadruplex-cross-linked catalytic hydrogel mimicking the function of horseradish peroxidase, reflected by the catalyzed oxidation of 2,2'-azinobis-(3-ethylbenzthiazoline-6-sulfonic acid), ABTS(2-), by H2O2 to ABTS(·-) and by the catalyzed generation of chemiluminescence in the presence of luminol/H2O2. Cyclic "ON" and "OFF" activation of the catalytic functions of the hydrogel are demonstrated upon the formation of the hydrogel in the presence of K(+) ions and its dissociation by 18-crown-6 ether, respectively. The hydrogel is characterized by rheology measurements, circular dichroism, and probing its chemical and photophysical properties.
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered