Merck
  • Pediatric anophthalmic sockets and orbital implants: outcomes with polymer-coated implants.
Pediatric anophthalmic sockets and orbital implants: outcomes with polymer-coated implants.
Ophthalmology (2013-02-13)
Maria Kirzhner, Yevgeniy Shildkrot, Barrett G Haik, Ibrahim Qaddoumi, Carlos Rodriguez-Galindo, Matthew W Wilson
ABSTRACT
To compare wrapped and polymer-coated hydroxyapatite implants in children undergoing primary enucleation with no adjuvant therapies. Retrospective, interventional cohort study. All children undergoing primary enucleation without adjuvant therapies between 1999 and 2009 at a tertiary pediatric cancer hospital. Review and analysis of patient records. Implant exposure, extrusion and migration, socket contracture, and formation of pyogenic granuloma. Sixty consecutive patients undergoing primary enucleation with no adjuvant chemotherapy or radiation with follow-up of at least 12 months were included. Retinoblastoma was the diagnosis in 59 eyes (98.3%). Median follow-up was 3.6 years (range, 1.0-9.3 years). Two implant sizes were used: 20 mm in 47 patients (78.3%) and 18 mm in 13 patients (21.7%). Overall, 52 patients (86.7%) had an event-free recovery. Polymer-coated hydroxyapatite implants (43/60, 71.7%), when compared with wrapped ones (17/60, 28.3%), had a trend toward greater event-free recovery (odds ratio [OR], 1.6; 95% confidence interval [CI], 0.3-7.7) and lower exposure rate (OR, 2.1; 95% CI, 0.4-10.5). The use of polymer-coated hydroxyapatite implants is associated with favorable outcomes in the pediatric population. Despite observed complications, long-term implant retention is possible in most children. The author(s) have no proprietary or commercial interest in any materials discussed in this article.
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000