Merck
A six-carbon 10π-electron aromatic system supported by group 3 metals.
Nature communications (2013-02-07)
Wenliang Huang, Florian Dulong, Tianpin Wu, Saeed I Khan, Jeffrey T Miller, Thibault Cantat, Paula L Diaconescu
ABSTRACT
Aromaticity is a fundamental concept with implications spanning all the chemical sciences. Hückel's (4n+2)π-electron rule is the standard criterion to determine aromaticity and it applies well to neutral arenes as well as to charged species such as the cyclopentadienyl anion, the cyclooctatetraene dianion and the cycloheptatrienyl cation (tropylium). In the series of all-carbon aromatic compounds, no example of a benzene tetraanion, which is theoretically a 6C, 10π-electron aromatic system, has been reported although heteroatom analogues of such a system, known as 'electron-rich aromatics', have been studied in detail for a long time. Here we present the isolation of the first tetraanionic-substituted benzene as a ligand coordinated to group 3 metals. The nature of the substituted benzene tetraanion and the aromaticity of the 6C, 10π-electron system were established by X-ray crystallographic studies, multi-nuclei nuclear magnetic resonance spectroscopy, X-ray absorption spectroscopy and density functional theory calculations.
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol