Merck
Single-layer single-crystalline SnSe nanosheets.
Journal of the American Chemical Society (2013-01-15)
Lun Li, Zhong Chen, Ying Hu, Xuewen Wang, Ting Zhang, Wei Chen, Qiangbin Wang
ABSTRACT
Single-layer single-crystalline SnSe nanosheet with four-atomic thickness of ~1.0 nm and lateral size of ~300 nm is presented here by using a one-pot synthetic method. It is found that 1,10-phenanthroline plays an important role in determining the morphology of the SnSe product as three-dimensional SnSe nanoflowers are obtained in the absence of 1,10-phenanthroline while keeping other reaction parameters the same. The evolution process study discloses that single-crystalline nanosheets are obtained from the coalescence of the SnSe nucleus in an orientated attachment mechanism. Band gap determination and optoelectronic test based on hybrid films of SnSe and poly(3-hexylthiophene) indicate the great potential of the ultrathin SnSe nanosheets in photodector and photovoltaic, and so forth.
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm