Merck
  • A conceptual translation of homogeneous catalysis into heterogeneous catalysis: homogeneous-like heterogeneous gold nanoparticle catalyst induced by ceria supporter.
A conceptual translation of homogeneous catalysis into heterogeneous catalysis: homogeneous-like heterogeneous gold nanoparticle catalyst induced by ceria supporter.
Nanoscale (2013-01-10)
Zhen-Xing Li, Wei Xue, Bing-Tao Guan, Fu-Bo Shi, Zhang-Jie Shi, Hong Jiang, Chun-Hua Yan
ABSTRACT
Translation of homogeneous catalysis into heterogeneous catalysis is a promising solution to green and sustainable development in chemical industry. For this purpose, noble metal nanoparticles represent a new frontier in catalytic transformations. Many challenges remain for researchers to transform noble metal nanoparticles of heterogeneous catalytic active sites into ionic species of homogeneous catalytic active sites. We report here a successful design on translating homogeneous gold catalysis into a heterogeneous system with a clear understanding of the catalytic pathway. This study initiates a novel concept to immobilize a homogeneous catalyst based on electron transfer between supporting base and supported nanoparticles. Meanwhile, on the basis of theoretical calculation, it has deepened the understanding of the interactions between noble metal nanoparticles and the catalyst support.
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O