Merck
  • A comparison of glucose concentration in paired specimens collected in serum separator and fluoride/potassium oxalate blood collection tubes under survey 'field' conditions.
A comparison of glucose concentration in paired specimens collected in serum separator and fluoride/potassium oxalate blood collection tubes under survey 'field' conditions.
Clinical biochemistry (2012-12-12)
Lois Fernandez, Penny Jee, Mari-Jill Klein, Peter Fischer, Sherry L Perkins, Stephen P J Brooks
ABSTRACT
There are no direct comparisons of blood glucose values in samples collected with barrier serum tubes (SST) and NaF/potassium oxalate (NaF/KOx) plasma tubes. Collection of samples in SST tubes can offer considerable savings and specimen processing advantages for national level surveys. Serum and plasma samples were collected under 'field conditions' from a single draw of 3692 individuals participating in the Canadian Health Measures Survey. The samples were analyzed retrospectively using the VITROS GLU Slide method (glucose oxidase-based). There was a high rate of hemolysis in the NaF/KOx tubes (86.2%) while hemolysis was infrequently observed with the SST tubes (2%). Comparing only blood draws where no hemolysis was observed in both tubes (n=495; paired t-test) showed no effect of tube type on serum/plasma glucose concentrations. This was also observed when data was restricted to cases when only SST samples were not hemolyzed (n=3546; paired t-test). These data show that both collection tubes can be used under survey collection and processing conditions to measure glucose with our assay system with no difference in reported results.
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid solution, 0.05 M
Sigma-Aldrich
Oxalic acid solution, 0.5 M
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, SAJ first grade, ≥97.0%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate solution, 0.0125 M
Sigma-Aldrich
Sodium oxalate solution, 5 mM