Merck
  • Pd-catalyzed β-selective direct C-H bond arylation of thiophenes with aryltrimethylsilanes.
Pd-catalyzed β-selective direct C-H bond arylation of thiophenes with aryltrimethylsilanes.
Organic letters (2012-12-12)
Kenji Funaki, Tetsuo Sato, Shuichi Oi
ABSTRACT
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, SAJ first grade, ≥98.0%
Sigma-Aldrich
Copper(II) chloride, powder, 99%