Merck
  • Impact of treatment on Pb release from full and partially replaced harvested Lead Service Lines (LSLs).
Impact of treatment on Pb release from full and partially replaced harvested Lead Service Lines (LSLs).
Water research (2012-11-24)
Clément Cartier, Evelyne Doré, Laurent Laroche, Shokoufeh Nour, Marc Edwards, Michèle Prévost
ABSTRACT
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the relatively short duration partial replacement of lead pipe by copper pipe (3 months), generated high lead release, attributed to galvanic corrosion, resulting in a final outcome for lead release that was even worse than for a full lead pipe. Increased lead release was especially evident at higher flow rates. Orthophosphate reduced lead release from full lead pipes by 64%. For partially replaced samples with copper, lead concentrations were unchanged by phosphate dosing at moderate flow (103 ± 265 vs 169 ± 349 μg/L) and were increased to very high levels when sampled at high flow rates (1001 ± 1808 vs 257 ± 224 μg/L). The increase lead release was in the form of particulate lead (>90%). In comparison to the condition without treatment, increased sulfate treatment had little impact on lead release from 100%-Pb rigs but reduced lead release from partially replaced lead pipes with copper. Our results also raise questions concerning protocols based on short 30 min stagnation (as those used in Canada) due to their incapacity to consider particulate lead release generated mostly after longer stagnation.
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium sulfate decahydrate, reagent grade, 97%