Merck
  • Protective effects of polydatin from Polygonum cuspidatum against carbon tetrachloride-induced liver injury in mice.
Protective effects of polydatin from Polygonum cuspidatum against carbon tetrachloride-induced liver injury in mice.
PloS one (2012-10-03)
Hong Zhang, Cheng-Hao Yu, Yi-Ping Jiang, Cheng Peng, Kun He, Jian-Yuan Tang, Hai-Liang Xin
ABSTRACT
Polydatin is one of main compounds in Polygonum cuspidatum, a plant with both medicinal and nutritional value. The possible hepatoprotective effects of polydatin on acute liver injury mice induced by carbon tetrachloride (CCl(4)) and the mechanisms involved were investigated. Intraperitoneal injection of CCl(4) (50 Āµl/kg) resulted in a significant increase in the levels of serum aspartate aminotransferase (AST), alanine aminotransferase (ALT) and hepatic malondialdehyde (MDA), also a marked enhancement in the expression of hepatic tumor necrosis factor-alpha (TNF-Ī±), interleukin-1 beta (IL-1Ī²), cyclooxygenase-2 (COX-2), inducible nitric oxide synthase (iNOS) and nuclearfactor-kappa B (NF-ĪŗB). On the other hand, decreased glutathione (GSH) content and activities of glutathione transferase (GST), superoxide dismutase (SOD), catalase (CAT) and glutathione peroxidase (GPx) were observed following CCl(4) exposure. Nevertheless, all of these phenotypes were evidently reversed by preadministration of polydatin for 5 continuous days. The mRNA and protein expression levels of hepatic growth factor-beta1 (TGF-Ī²(1)) were enhanced further by polydatin. These results suggest that polydatin protects mice against CCl(4)-induced liver injury through antioxidant stress and antiinflammatory effects. Polydatin may be an effective hepatoprotective agent and a promising candidate for the treatment of oxidative stress- and inflammation-related diseases.
Sigma-Aldrich
Polydatin, ≥95% (HPLC)