Merck
  • Impact of vitamin B12 on formation of the tetrachloroethene reductive dehalogenase in Desulfitobacterium hafniense strain Y51.
Impact of vitamin B12 on formation of the tetrachloroethene reductive dehalogenase in Desulfitobacterium hafniense strain Y51.
Applied and environmental microbiology (2012-09-11)
Anika Reinhold, Martin Westermann, Jana Seifert, Martin von Bergen, Torsten Schubert, Gabriele Diekert
ABSTRACT
Corrinoids are essential cofactors of reductive dehalogenases in anaerobic bacteria. Microorganisms mediating reductive dechlorination as part of their energy metabolism are either capable of de novo corrinoid biosynthesis (e.g., Desulfitobacterium spp.) or dependent on exogenous vitamin B(12) (e.g., Dehalococcoides spp.). In this study, the impact of exogenous vitamin B(12) (cyanocobalamin) and of tetrachloroethene (PCE) on the synthesis and the subcellular localization of the reductive PCE dehalogenase was investigated in the gram-positive Desulfitobacterium hafniense strain Y51, a bacterium able to synthesize corrinoids de novo. PCE-depleted cells grown for several subcultivation steps on fumarate as an alternative electron acceptor lost the tetrachloroethene-reductive dehalogenase (PceA) activity by the transposition of the pce gene cluster. In the absence of vitamin B(12), a gradual decrease of the PceA activity and protein amount was observed; after 5 subcultivation steps with 10% inoculum, more than 90% of the enzyme activity and of the PceA protein was lost. In the presence of vitamin B(12), a significant delay in the decrease of the PceA activity with an ∼90% loss after 20 subcultivation steps was observed. This corresponded to the decrease in the pceA gene level, indicating that exogenous vitamin B(12) hampered the transposition of the pce gene cluster. In the absence or presence of exogenous vitamin B(12), the intracellular corrinoid level decreased in fumarate-grown cells and the PceA precursor formed catalytically inactive, corrinoid-free multiprotein aggregates. The data indicate that exogenous vitamin B(12) is not incorporated into the PceA precursor, even though it affects the transposition of the pce gene cluster.
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Tetrachloroethylene, suitable for HPLC, ≥99.9%
Supelco
Tetrachloroethylene, analytical standard
Sigma-Aldrich
Tetrachloroethylene, ≥99.5%
Supelco
Density Standard 1623 kg/m3, H&D Fitzgerald Ltd. Quality