Merck
  • Hydrothermal reaction kinetics and pathways of phenylalanine alone and in binary mixtures.
Hydrothermal reaction kinetics and pathways of phenylalanine alone and in binary mixtures.
ChemSusChem (2012-08-29)
Shujauddin Changi, Minghan Zhu, Phillip E Savage
ABSTRACT
We examined the behavior of phenylalanine in high-temperature water (HTW) at 220, 250, 280, and 350 °C. Under these conditions, the major product is phenylethylamine. The minor products include styrene and phenylethanol (1-phenylethanol and 2-phenylethanol), which appear at higher temperatures and longer batch holding times. Phenylethylamine forms via decarboxylation of phenylalanine, styrene forms via deamination of phenylethylamine, and phenylethanol forms via hydration of styrene. We quantified the molar yields of each product at the four temperatures, and the carbon recovery was between 80-100 % for most cases. Phenylalanine disappearance follows first-order kinetics with an activation energy of 144 ± 14 kJ mol⁻¹ and a pre-exponential factor of 10(12.4 ± 1.4)  min⁻¹. A kinetics model based on the proposed pathways was consistent with the experimental data. Effects of five different salts (NaCl, NaNO₃, Na₂ SO₄, KCl, K₂ HPO₄) and boric acid (H₃BO₃) on phenylalanine behavior at 250 °C have also been elucidated. These additives increase phenylalanine conversion, but decrease the yield of phenylethylamine presumably by promoting formation of high molecular weight compounds. Lastly, binary mixtures of phenylalanine and ethyl oleate have been studied at 350 °C and three different molar concentration ratios. The presence of phenylalanine enhances the conversion of ethyl oleate and molar yields of fatty acid. Higher concentration of ethyl oleate leads to increased deamination of phenylethylamine and hydration of styrene. Amides are also formed due to the interaction of oleic acid/ethyl oleate and phenylethylamine/ammonia and lead to a decrease in the fatty acid yields. Taken collectively, these results provide new insights into the reactions of algae during its hydrothermal liquefaction to produce crude bio-oil.
Sigma-Aldrich
Ethyl oleate, technical grade, 70%
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl oleate, 98%
Sigma-Aldrich
Ethyl oleate, natural, ≥85%