Merck
  • Fatal manganese intoxication due to an error in the elaboration of Epsom salts for a liver cleansing diet.
Fatal manganese intoxication due to an error in the elaboration of Epsom salts for a liver cleansing diet.
Forensic science international (2012-08-14)
Baltasar Sánchez, Jaume Casalots-Casado, Salvador Quintana, Amparo Arroyo, Carles Martín-Fumadó, Ignasi Galtés
ABSTRACT
We describe the case of a 50-year-old man with a fatal intoxication after accidental massive oral ingestion of manganese. The patient presented with lethargy, diffuse abdominal pain, vomiting, and profuse diarrhea after ingesting Epsom salts (magnesium sulfate heptahydrate) during a liver cleansing diet. Despite intensive care management with intubation, prone position ventilation, continuous venovenous hemofiltration, and multiple transfusions, he progressed to refractory shock with multiple organ dysfunction resulting in death within 72 h. Similar patients arrived at several hospitals with identical epidemiology (all had ingested the same salt obtained in the same place). Clinical and forensic investigations (X-ray diffraction) discovered that the supplier had mistakenly prepared the salts with hydrated manganese sulfate instead of magnesium sulfate heptahydrate. The results enabled the other patients to be successfully treated for hydrated manganese sulfate intoxication with life support in the intensive care unit and chelation therapy (EDTA). We describe the clinical presentation of acute manganese poisoning and alert professionals to the risk of an increasingly popular diet. This case demonstrates the importance of collaboration between clinicians, pathologists, and forensic scientists to resolve a difficult-to-diagnose case.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) sulfate monohydrate, puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%