Merck
  • Total synthesis of aspercyclides A and B via intramolecular oxidative diaryl ether formation.
Total synthesis of aspercyclides A and B via intramolecular oxidative diaryl ether formation.
Organic letters (2012-08-09)
Tatsuya Yoshino, Itaru Sato, Masahiro Hirama
ABSTRACT
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Sigma-Aldrich
Diethyl ether, ≥99.5%
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free