Merck
  • Nanoscale manganese oxide within Faujasite zeolite as an efficient and biomimetic water oxidizing catalyst.
Nanoscale manganese oxide within Faujasite zeolite as an efficient and biomimetic water oxidizing catalyst.
Dalton transactions (Cambridge, England : 2003) (2012-07-27)
Mohammad Mahdi Najafpour, Babak Pashaei
ABSTRACT
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in the presence of cerium(IV) ammonium nitrate as a non-oxo transfer oxidant.
Sigma-Aldrich
Ammonium nitrate, SAJ first grade
Sigma-Aldrich
Ammonium nitrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, ≥99.0%
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%