Merck
  • [Selective staining of pericentromeric heterochromatin regions in chromosomes of spontaneously dividing cells with the use of the acridine orange fluorochrome].
[Selective staining of pericentromeric heterochromatin regions in chromosomes of spontaneously dividing cells with the use of the acridine orange fluorochrome].
Genetika (2012-06-27)
T V Kuznetzova, I L Trofimova, M S Liapunov, E V Evdokimenko, V S Baranov
ABSTRACT
A novel phenomenon of unusual selective acridine orange (AO) staining ofpericentromeric heterochromatin regions (HRs) in chromosomal preparations from tissue with known spontaneous mitotic activity (chorionic villi, placenta, embryonic tissues, bone marrow, and testes), as well as embryonic stem cells, is described. Staining with 0.01% AO in a citric-phosphate (pH 5.5) or sodium phosphate (pH 7.0) buffer solution allows the HRs of human chromosomes (1q12, 9q12, 13p11.2, 14p11.2, 15p11.2, 16q11.2, 21p11.2, 22p11.2, and Yq12) and pericentromeric HRs of mouse chromosomes to be reliably detected by the red fluorescence of AO. This method of AO staining does not require any pretreatment. Explanations for metachromatic AO staining of polymorphic pericentromeric HRs in chromosomes of spontaneously dividing cells are suggested. A high reproducibility of the specific AO staining makes it possible to suggest its use as a reliable quick method for detection of polymorphic HRs of human chromosomes in cytogenetic prenatal diagnosis and oncohematology.
Sigma-Aldrich
Acridine Orange solution, 2% in H2O
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Acridine Orange base, Dye content 75 %