Merck
  • Effects of laccase on lignin depolymerization and enzymatic hydrolysis of ensiled corn stover.
Effects of laccase on lignin depolymerization and enzymatic hydrolysis of ensiled corn stover.
Bioresource technology (2012-05-23)
Qin Chen, Megan N Marshall, Scott M Geib, Ming Tien, Tom L Richard
ABSTRACT
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS) analysis. The significant findings suggest that ensilage might provide a platform for biological pretreatment. By partially hydrolyzing cellulose and hemicellulose into soluble sugars, ensilage facilitates laccase penetration into the lignocellulose complex to enhance lignin degradation. Downstream cellulose hydrolysis was improved 7% with increasing laccase loading rate. These results demonstrate the potential of enzymes, either directly amended or expressed by microbes during ensilage, to maximize utilization of corn stover for cellulosic biofuels and other downstream fermentations.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium chloride solution
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)