Merck
  • Enhancement of the nanofibrillation of wood cellulose through sequential periodate-chlorite oxidation.
Enhancement of the nanofibrillation of wood cellulose through sequential periodate-chlorite oxidation.
Biomacromolecules (2012-04-20)
Henrikki Liimatainen, Miikka Visanko, Juho Antti Sirviö, Osmo E O Hormi, Jouko Niinimaki
ABSTRACT
Sequential regioselective periodate-chlorite oxidation was employed as a new and efficient pretreatment to enhance the nanofibrillation of hardwood cellulose pulp through homogenization. The oxidized celluloses with carboxyl contents ranging from 0.38 to 1.75 mmol/g could nanofibrillate to highly viscous and transparent gels with yields of 100-85% without clogging the homogenizer (one to four passes). On the basis of field-emission scanning electron microscopy images, the nanofibrils obtained were of typical widths of approximately 25 ± 6 nm. All of the nanofibrillar samples maintained their cellulose I crystalline structure according to wide-angle X-ray diffraction results, and the crystallinity index was approximately 40% for all samples.
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%