Merck
  • Substrate mass transport in two-phase partitioning bioreactors employing liquid and solid non-aqueous phases.
Substrate mass transport in two-phase partitioning bioreactors employing liquid and solid non-aqueous phases.
Bioprocess and biosystems engineering (2012-03-29)
Hala Fam, Andrew J Daugulis
ABSTRACT
The mass transfer of phenol and butyl acetate to/from water was studied in two-phase partitioning bioreactors using immiscible organic solvents and solid polymer beads as the partitioning phases in a 5-L stirred tank bioreactor. Virtually instantaneous mass transfer was observed with phenol in water/2-undecanone, and with butyl acetate in water/silicone oil systems. The mass transfer of butyl acetate to silicone oil was rapid irrespective of the viscosity of the partitioning phase. When Hytrel® polymer beads were employed as the partitioning phase, substrate transport to the polymer was found not to be externally mass transfer limited, but rather internally by substrate diffusion into the polymer. In contrast to gaseous, poorly soluble substrates studied in other works, mass transfer of soluble substrates such as phenol and butyl acetate to the polymer was unaffected by impeller speed but rather by polymer mass fraction.
Sigma-Aldrich
Butyl acetate, ≥99.5%, suitable for atomic absorption spectrometry
Sigma-Aldrich
Butyl acetate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Butyl acetate, JIS special grade, ≥99.0%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%