Merck
  • Incorporation of initiation, promotion and inhibition in the Rct concept and its application in determining the initiation and inhibition capacities of natural water in ozonation.
Incorporation of initiation, promotion and inhibition in the Rct concept and its application in determining the initiation and inhibition capacities of natural water in ozonation.
Water research (2012-02-14)
Ee Ling Yong, Yi-Pin Lin
ABSTRACT
Ozonation is widely employed in water treatment to purify water. The R(ct) concept, which is defined as the ratio of OH exposure to ozone exposure, has been commonly used to quantify the OH concentration generating from ozone decomposition and model the removal of organic pollutant. Due to its empirical nature, however, the R(ct) concept is limited in quantitatively elucidating how initiator, promoter and inhibitor involved in the OH chain reactions affect its value. A new R(ct) model was developed by integrating the R(ct) concept and the transient steady-state OH concentration to evaluate the influences of these modes of reactions on the R(ct) value. It was found that the R(ct) value is not only the ratio of OH exposure to ozone exposure but also the ratio of the total initiation capacity to the total inhibition capacity in a system. The presence of promoter, however, does not affect the R(ct) value although it does accelerate ozone decomposition leading to lower ozone and OH exposures and result in hindered removal of target pollutant. The hindered removal of ibuprofen by ozonation in the presence of methanol (promoter) can be quantitatively described by the new R(ct) model. The model can also be used to quantify the initiation and inhibition capacities of an ozonation system via the addition of an external inhibitor. Its application in determining the initiation and inhibition capacities of natural water was demonstrated.
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
tert-Butanol, SAJ first grade, ≥98.0%
Sigma-Aldrich
tert-Butanol, SAJ special grade, ≥99.0%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)