Merck
  • Gd₂O₃ nanoparticles in hematopoietic cells for MRI contrast enhancement.
Gd₂O₃ nanoparticles in hematopoietic cells for MRI contrast enhancement.
International journal of nanomedicine (2012-01-10)
Anna Hedlund, Maria Ahrén, Håkan Gustafsson, Natalia Abrikossova, Marcel Warntjes, Jan-Ingvar Jönsson, Kajsa Uvdal, Maria Engström
ABSTRACT
As the utility of magnetic resonance imaging (MRI) broadens, the importance of having specific and efficient contrast agents increases and in recent time there has been a huge development in the fields of molecular imaging and intracellular markers. Previous studies have shown that gadolinium oxide (Gd(2)O(3)) nanoparticles generate higher relaxivity than currently available Gd chelates: In addition, the Gd(2)O(3) nanoparticles have promising properties for MRI cell tracking. The aim of the present work was to study cell labeling with Gd(2)O(3) nanoparticles in hematopoietic cells and to improve techniques for monitoring hematopoietic stem cell migration by MRI. Particle uptake was studied in two cell lines: the hematopoietic progenitor cell line Ba/F3 and the monocytic cell line THP-1. Cells were incubated with Gd(2)O(3) nanoparticles and it was investigated whether the transfection agent protamine sulfate increased the particle uptake. Treated cells were examined by electron microscopy and MRI, and analyzed for particle content by inductively coupled plasma sector field mass spectrometry. Results showed that particles were intracellular, however, sparsely in Ba/F3. The relaxation times were shortened with increasing particle concentration. Relaxivities, r(1) and r(2) at 1.5 T and 21°C, for Gd(2)O(3) nanoparticles in different cell samples were 3.6-5.3 s(-1) mM(-1) and 9.6-17.2 s(-1) mM(-1), respectively. Protamine sulfate treatment increased the uptake in both Ba/F3 cells and THP-1 cells. However, the increased uptake did not increase the relaxation rate for THP-1 as for Ba/F3, probably due to aggregation and/or saturation effects. Viability of treated cells was not significantly decreased and thus, it was concluded that the use of Gd(2)O(3) nanoparticles is suitable for this type of cell labeling by means of detecting and monitoring hematopoietic cells. In conclusion, Gd(2)O(3) nanoparticles are a promising material to achieve positive intracellular MRI contrast; however, further particle development needs to be performed.
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%