Merck
  • Self-assembled mesoporous γ-Al2O3 spherical nanoparticles and their efficiency for the removal of arsenic from water.
Self-assembled mesoporous γ-Al2O3 spherical nanoparticles and their efficiency for the removal of arsenic from water.
Journal of hazardous materials (2011-12-16)
Astam K Patra, Arghya Dutta, Asim Bhaumik
ABSTRACT
We report a highly efficient synthetic strategy for self-assembled mesoporous γ-Al(2)O(3) materials using sodium salicylate as template. The mesoporous γ-Al(2)O(3) samples synthesized following this strategy have high surface areas (231-497 m(2)g(-1)), consist of crystalline tiny spherical nanoparticles of dimensions ca. 2-10nm and showed high affinity for the adsorption of arsenic from the contaminated aqueous solutions. Efficient synthesis strategy, exceptionally high surface area and high adsorption efficiency of these mesoporous γ-Al(2)O(3) materials for the dissolved arsenic from the contaminated aqueous solutions (in the form of oxyanions of arsenic) could find potential utility in the purification of polluted water.
Sigma-Aldrich
Sodium salicylate, SAJ first grade, ≥99.5%
Sigma-Aldrich
Sodium salicylate, puriss. p.a., reag. Ph. Eur., 99.5-101.0% (calc. to the dried substance)
Sigma-Aldrich
Sodium salicylate, ≥99.5% (HPLC), puriss. p.a.
Sigma-Aldrich
Sodium salicylate, 98.0-102.0% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Sodium salicylate, ReagentPlus®, ≥99.5% (titration)