Merck
  • Use of the RapID-ANA system and sodium polyanetholesulfonate disk susceptibility testing in identifying Haemophilus ducreyi.
Use of the RapID-ANA system and sodium polyanetholesulfonate disk susceptibility testing in identifying Haemophilus ducreyi.
Journal of clinical microbiology (1990-01-01)
R Shawar, J Sepulveda, J E Clarridge
ABSTRACT
Haemophilus ducreyi has traditionally been difficult to identify. We have utilized simple test methods to identify 19 fresh isolates obtained during a recent outbreak of chancroid in Houston and six strains of H. ducreyi from other outbreaks. Tests were performed from growth on chocolate agar after 48 h of incubation at 35 degrees C with increased humidity and CO2. All isolates exhibited typical colonial morphology and Gram stain. Isolates were catalase negative and oxidase and nitrate positive (in enriched broth). The RapID NH system failed to identify these strains because of negative reactions with alkaline phosphatase and nitrate reductase. However, by using the RapID-ANA system, all strains were positive for alkaline phosphatase and arginine, glycine, and serine aminopeptidases. Their biochemical profiles were distinct from those obtained with 66 strains representing 13 species similar to H. ducreyi. We also investigated the use of sodium polyanetholesulfonate (SPS) disk susceptibility to identify and differentiate H. ducreyi from other species. All H. ducreyi isolates were susceptible, as evidenced by the presence of a zone of inhibition with an average size of 15 mm around the SPS disk. With the exceptions of Neisseria gonorrhoeae, Gardnerella vaginalis, and Capnocytophaga spp., no other strain showed any evidence of inhibition. The latter three organisms can be easily differentiated from H. ducreyi by various features including reactions in the RapID-ANA. We conclude that, by considering simple growth and biochemical characteristics, SPS susceptibilities, and reactions in RapID-ANA, it is possible for more clinical laboratories to definitively identify this organism.
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, powder
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, BioUltra