Merck
  • Aptamer-mediated surface-enhanced Raman spectroscopy intensity amplification.
Aptamer-mediated surface-enhanced Raman spectroscopy intensity amplification.
Nano letters (2010-09-25)
Nam Hoon Kim, Seung Joon Lee, Martin Moskovits
ABSTRACT
A bifunctional adenosine-sensitive double-stranded DNA aptamer was used to create and control a surface-enhanced Raman spectroscopy (SERS) hot spot between a bulk Au surface and a gold nanoparticle (Au NP) attached to the aptamer via a biotin-avidin linkage. The Au NP was decorated with 4-aminobenzenethiol (4-ABT), a Raman reporter molecule. In the presence of adenosine, the target molecule, the SERS spectrum of 4-ABT increased in intensity by (concentration-dependent) factors as large as ∼4; in situ, atomic force microscopy imaging showed the mean height of the Au NP-bearing aptamer to decrease by ∼5 nm consistent with the observed SERS intensity change. Because the aptamer's geometrical change is induced by one or two molecules, while the resulting SERS intensity changes involve many reporter molecules residing in the modified hot spot, the aptamer amplifies the SERS effect.
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Sulfanilic acid, JIS special grade, 99.0-100.5%
Millipore
Nitrate Reagent B, suitable for microbiology