Merck
Synthesis of nanometre-thick MoO3 sheets.
Nanoscale (2010-07-21)
Kourosh Kalantar-zadeh, Jianshi Tang, Minsheng Wang, Kang L Wang, Alexandros Shailos, Kosmas Galatsis, Robert Kojima, Veronica Strong, Andrew Lech, Wojtek Wlodarski, Richard B Kaner
ABSTRACT
The formation of MoO(3) sheets of nanoscale thickness is described. They are made from several fundamental sheets of orthorhombic alpha-MoO(3), which can be processed in large quantities via a low cost synthesis route that combines thermal evaporation and mechanical exfoliation. These fundamental sheets consist of double-layers of linked distorted MoO(6) octahedra. Atomic force microscopy (AFM) measurements show that the minimum resolvable thickness of these sheets is 1.4 nm which is equivalent to the thickness of two double-layers within one unit cell of the alpha-MoO(3) crystal.
Sigma-Aldrich
Molybdenum(VI) oxide, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, nanopowder, 100 nm (TEM), 99.5% trace metals basis
Sigma-Aldrich
Molybdenum(VI) oxide, ACS reagent, ≥99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, puriss. p.a., 99.5%
Sigma-Aldrich
Molybdenum(VI) oxide, ReagentPlus®, ≥99.5%