Merck
  • Analysis of adulterated herbal medicines and dietary supplements marketed for weight loss by DOSY 1H-NMR.
Analysis of adulterated herbal medicines and dietary supplements marketed for weight loss by DOSY 1H-NMR.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2010-05-04)
J Vaysse, S Balayssac, V Gilard, D Desoubdzanne, M Malet-Martino, R Martino
ABSTRACT
Twenty herbal medicines or dietary supplements marketed as natural slimming products were analysed by diffusion ordered spectroscopy (DOSY) 1H-nuclear magnetic resonance (NMR) and DOSY-COSY 1H-NMR. The method allows analysis of the whole sample with the detection of both active and inactive ingredients in these complex matrices. Among the 20 formulations analysed, two were strictly herbal and four had a composition corresponding to declared ingredients on the packaging or the leaflet. The others were all adulterated. Eight formulations contain sibutramine alone at doses ranging from 4.4 to 30.5 mg/capsule. Five formulations contain sibutramine (from 5.0 to 19.6 mg/capsule or tablet) in combination with phenolphthalein (from 4.4 to 66.1 mg/capsule), and the last formulation was adulterated with synephrine (19.5 mg/capsule). Quantification of the actives was carried out with 1H-NMR. Several other compounds were also characterized including methylsynephrine, vitaberin, sugars, vitamins, etc. DOSY NMR is thus proposed as a useful tool for detection of unexpected adulteration.
Sigma-Aldrich
Phenolphthalein, SAJ first grade, ≥97.0%
Sigma-Aldrich
Phenolphthalein, JIS special grade, ≥98.0%
Sigma-Aldrich
Phenolphthalein solution, 0.04 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein solution, 0.1 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein solution, 1 % (w/v), pH 7.8-10.0
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)