Merck
The effects of lead and zinc on the quality of semen of albino rats.
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP (2009-08-05)
Muhammad Rafique, Naheed Khan, Khalida Perveen, Anjum Naqvi
ABSTRACT
To determine the effects of lead and zinc administration on the quality of semen of albino rats. Experimental study. Basic Medical Sciences Institute, Jinnah Postgraduate Medical Centre, Karachi, from August 2003 to December 2005. Sixty adult albino rats selected for the study were divided into three groups, group A received injection normal saline 1 ml intraperitoneally daily for 8 weeks. Group B received lead chloride in a dose of 10 mg/kg body weight intraperitoneally daily. Group C received lead chloride in a dose of 10 mg/kg body weight and zinc chloride in a dose of 1 mg/kg body weight intraperitoneally daily. On the day of completion of treatment, the animals were sacrificed; epididymis was used for semen analysis. Student's t-test was used to determinate significance; the p-value < 0.05 was taken as significant. The number of sperms was 7.3, 1.7 and 6.6 million cells/ml in groups A, B and C respectively. Sperm's count decreased by 87% in group B (p < 0.001, CI 4117082.4 - 6965747.6) as compared to group A. Compared with group C, the sperm's count was decreased to 75% (p < 0.001, CI -5417413 to -4416987). The immotility of sperms was 27%, 57% and 26% in groups A, B and C respectively. There was 30% decreased motility of sperm in group B (p < 0.001, CI -30.19425 to -19.80575) as compared to group A. Compared with group C, the immotile sperm were increased to 31% (p < 0.001, CI 19.87494 - 30.92506). Lead produced toxic effects on germinal epithelium and altered the quality of semen which was improved by zinc.
Sigma-Aldrich
Lead(II) chloride, AnhydroBeads, −10 mesh, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Lead(II) chloride, powder, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Lead(II) chloride, 99.999% trace metals basis
Pricing and availability is not currently available.