Merck
  • Characterisation of mixed lithium dodecyl sulphate/lithium perfluorooctanesulphonate pseudo-stationary phases in MEKC.
Characterisation of mixed lithium dodecyl sulphate/lithium perfluorooctanesulphonate pseudo-stationary phases in MEKC.
Electrophoresis (2009-06-12)
András Varga, Mónika Huszár, Zsófia Dobos, Eva Kiss, Anikó Horváth, Miklós Idei
ABSTRACT
Lipophilicity and methylene selectivity of mixed pseudo-stationary phases (PSPs) (containing lithium dodecyl sulphate (LDS) and lithium perfluorooctanesulphonate (LiPFOS) in different molar ratios) applied in MEKC have been investigated. Micellar proportion (t(prop,mic), a quantity expressing that how much time is spent by the analyte in the micellar phase related to its whole migration time), CLOGP(50) value (showing the value of hydrophobicity of a molecule spending exactly 50% of its migration time in the PSP) and methylene selectivity have been determined for different LDS/LiPFOS mixed phases. Values of the above-mentioned parameters have been determined for analytes with different chemical structures (alkylbenzene and alkylphenone homologous series, alcohols). Good linear correlation was obtained between either the micellar proportion, CLOGP(50), or methylene selectivity and the phase composition for the mixed phases. Lipophilicity and methylene selectivity of the mixed LDS/LiPFOS PSPs can be calculated and can continuously be changed by mixing the two single phases (LDS and LiPFOS) in the appropriate (and calculable) portion.
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis