Merck
  • RNA extraction from various recalcitrant plant tissues with a cethyltrimethylammonium bromide-containing buffer followed by an acid guanidium thiocyanate-phenol-chloroform treatment.
RNA extraction from various recalcitrant plant tissues with a cethyltrimethylammonium bromide-containing buffer followed by an acid guanidium thiocyanate-phenol-chloroform treatment.
Bioscience, biotechnology, and biochemistry (2008-07-08)
Yuji Suzuki, Tadahiko Mae, Amane Makino
ABSTRACT
High-quality total RNA was extracted using a cethyltrimethylammonium bromide-containing buffer followed by an acid guanidium thiocyanate-phenol-chloroform treatment from recalcitrant plant tissues such as tree leaves (pine, Norway spruce, ginkgo, Japanese cedar, rose), flowers (rose, Lotus japonicus) and storage tissues (seeds of Lotus japonicus and rice, sweet potato tuber, banana fruit). This protocol greatly reduced the time required for RNA extraction.
Sigma-Aldrich
Guanidine thiocyanate solution, BioUltra, for molecular biology, ~6 M in H2O
Sigma-Aldrich
Guanidine thiocyanate, BioReagent, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine thiocyanate, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, for molecular biology, ≥99.0% (AT)