Merck
  • Blue-jean button nickel: prevalence and prevention of its release from buttons.
Blue-jean button nickel: prevalence and prevention of its release from buttons.
Dermatitis : contact, atopic, occupational, drug (2007-11-21)
Tina Suneja, Katherine H Flanagan, Dee Anna Glaser
ABSTRACT
Nickel is a common cause of allergic contact dermatitis and is associated with metal buttons and snaps on blue jeans. To determine the prevalence of nickel-positive metal buttons on blue jeans and the efficacy of nail polish in the prevention of nickel release. We collected 62 pairs of new and preworn blue jeans. The metal buttons were tested for nickel with a dimethylglyoxime test kit, and one coat of clear nail polish was applied to nickel-positive buttons. Painted buttons were dried for 24 hours, and the jeans were washed on delicate cycle and retested for nickel. The jeans underwent a second wash/dry cycle and were retested. The prevalence of nickel-positive metal buttons on blue jeans was 16%. Ten pairs of jeans tested positive for nickel. All 10 pairs tested negative for nickel after the application of one coat of clear nail polish. Of these 10 pairs, seven pairs underwent two wash/dry cycles, and all seven tested negative for nickel after each cycle. The presence of nickel-positive blue-jean buttons is greater in new jeans than in preworn jeans. One clear coat of nail polish can prevent nickel release through two wash/dry cycles in a test environment.
Sigma-Aldrich
Dimethylglyoxime, ≥97.0% (TLC)
Sigma-Aldrich
Dimethylglyoxime, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylglyoxime, puriss. p.a., ACS reagent, for the detection of Ni, ≥99.0% (TLC)
Sigma-Aldrich
Dimethylglyoxime, JIS special grade, ≥98.0%