Merck
  • A simple and rapid spectrophotometric method for the determination of nitrite by its decolorizing effect on peroxovanadate complex.
A simple and rapid spectrophotometric method for the determination of nitrite by its decolorizing effect on peroxovanadate complex.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-08-22)
Mansour S Abdul Galil, Mahadevaiah, M S Yogendra Kumar, G Nagendrappa
ABSTRACT
A rapid, simple, and most economical spectrophotometric method was proposed for the determination of nitrite in various water samples, soil samples, and roots of leguminous plants. The method is based on decolorizing effect of nitrite on complex formed between hydrogen peroxide and vanadate in acidic medium. The decolorization of that complex by nitrite was exploited to monitor the reaction spectrophotometrically at 470 nm. The method was optimized for effect of concentrations of ammonium metavanadate, hydrogen peroxide, various acids, concentrations of sulphuric acid, order of reagents addition and color stability. The color of the complex was found to be stable for about 2 days, and the stability constant of the complex was also calculated by modified Job's method. The linearity range of the calibration graph was over 6.67-66.7 microg ml(-1) of nitrite with molar absorptivity, 0.276 x 10(3) mol(-1) l cm(-1) and Sandell's sensitivity, 0.1667 microg cm(-2). The method was applied successfully for the determination of nitrite in soil samples, various wastewater samples and roots of leguminous plants.
Sigma-Aldrich
Ammonium metavanadate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Ammonium metavanadate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium metavanadate, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium metavanadate, 99.95% trace metals basis
Sigma-Aldrich
Ammonium metavanadate, 99%