Merck
  • p-Dimethylaminobenzaldehyde-reactive substances in tail tendon collagen of streptozotocin-diabetic rats: temporal relation to biomechanical properties and advanced glycation endproduct (AGE)-related fluorescence.
p-Dimethylaminobenzaldehyde-reactive substances in tail tendon collagen of streptozotocin-diabetic rats: temporal relation to biomechanical properties and advanced glycation endproduct (AGE)-related fluorescence.
Biochimica et biophysica acta (2000-11-09)
M Stefek, A Gajdosik, A Gajdosikova, L Krizanova
ABSTRACT
In the present work, pepsin digests of tail tendons from streptozotocin-diabetic rats were found to contain material that reacted rapidly at room temperature with p-dimethylaminobenzaldehyde (Ehrlich's reagent) to give an adduct with an absorbance spectrum characteristic of the Ehrlich chromogen of pyrrolic nature determined in ageing collagens. A significant correlation of the Ehrlich adduct with tendon mechanical strength and collagen fluorescence characteristic of advanced glycation endproducts was observed. Collagen content of the Ehrlich-positive material was found to be significantly elevated in tendons of diabetic rats compared with age-matched healthy controls. The results indicate that the p-dimethylaminobenzaldehyde-reactive pyrrole moieties may contribute to the increased cross-linking of diabetic matrix collagen. Profound inhibitory effect of aminoguanidine was observed, underlining the role of non-enzymatic mechanisms of advanced glycation in pyrrolisation and cross-linking of collagen exposed to hyperglycaemia. It is hypothesised that quantification of the p-dimethylaminobenzaldehyde-reactive material in matrix collagen may provide a tissue measure of integrated hyperglycaemia over prolonged periods of time. Further research is to assess the significance of p-dimethylaminobenzaldehyde-reactive substances in diabetic collagen tissues and to reveal their relationship to enzyme-mediated physiological pyrrolisation of ageing collagens.
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Supelco
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, JIS special grade, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Millipore
Kovac′s reagent for indoles, suitable for microbiology