Merck
  • [Effects of naringin on proliferation, differentiation and maturation of rat calvarial osteoblasts in vitro].
[Effects of naringin on proliferation, differentiation and maturation of rat calvarial osteoblasts in vitro].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-04-20)
Yuan-Kun Zhai, Yin-Bo Niu, Ya-Lei Pan, Chen-Rui Li, Xiang-Long Wu, Qi-Bing Mei
ABSTRACT
To investigate the effects of naringin on the proliferation, differention and maturaion of rat calvarial osteoblasts (ROB). Segregated neonatal SD rat skull, enzyme digestion to obtain ROB. The culture medium was replaced every three days. Serial subcultivation proceeded when cells covered with 80% culture dish. Naringin supplemented into the culture at 1 x 10(-4), 1 x 10(-5), 1 x 10(-6), 1 x 10(-7) mol x L(-1) respectively. MTT method was adopted in proliferation analysis and the activity of ALP was examined after induced 9 days. Search the best concentration and supplemented into the medium, then the osteogenic differentiation markers including the secretion amount of osteocalcin, osteopontin and bone morphogenetic protein-2 were compared between the naringin-supplemented group and the control. Total RNA was isolated and the mRNA level of bFGF, IGF-1, Runx-2, Osterix, ERa and ERbeta was investigated by Real time RT-PCR. Total protein also was isolated and the expression ERa, ERbeta and collagen I was examined by Western blot. After the addition of ICI 182.780, an inhibitor of the estrogen signal pathway, these index also was examined and the changes were compared. The ROB proliferation was motivated by naringin dose-dependently. And it evidently leads to osteogenic process and maturation. 1 x 10(-5) mol x L(-1) is the best concentration. Naringin improved the secretion of osteocalcin, osteopontin, bone morphogenetic protein-2 and collagen I significantly. Besides, it can also enhanced the mRNA level of bFGF, IGF-1, Runx-2, Osterix, ERalpha and ERbeta. While all these effects can be restrained by ICI 182.780. The naringin with final concentration of 1 x 10(-5) mol x L(-1) enhances the osteogenic differentiation and maturation of ROB significantly, while the promoting effects vanished after the addition of ICI 182.780. These results suggesting that naringin is one of the phytoestrogens and have the activity of bone formation may via estrogen signal pathway, it can be developed into a new drug for osteoporosis therapy.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Supelco
Naringin, analytical standard
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein