Merck
  • Mechanical induction of PGE2 in osteocytes blocks glucocorticoid-induced apoptosis through both the β-catenin and PKA pathways.
Mechanical induction of PGE2 in osteocytes blocks glucocorticoid-induced apoptosis through both the β-catenin and PKA pathways.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2010-06-26)
Yukiko Kitase, Leonardo Barragan, Hai Qing, Shino Kondoh, Jean X Jiang, Mark L Johnson, Lynda F Bonewald
ABSTRACT
Glucocorticoids are known to induce osteocyte apoptosis, whereas mechanical loading has been shown to sustain osteocyte viability. Here we show that mechanical loading in the form of fluid-flow shear stress blocks dexamethasone-induced apoptosis of osteocyte-like cells (MLO-Y4). Prostaglandin E(2) (PGE(2) ), a rapidly induced signaling molecule produced by osteocytes, was shown to be protective against dexamethasone-induced apoptosis, whereas indomethacin reversed the antiapoptotic effects of shear stress. This protective effect of shear stress was mediated through EP2 and EP4 receptors, leading to activation of the cAMP/protein kinase A signaling pathway. Activation of phosphatidylinositol 3-kinase, an inhibitor of glycogen synthesis kinase 3, also occurred, leading to the nuclear translocation of β-catenin, an important signal transducer of the Wnt signaling pathway. Both shear stress and prostaglandin increased the phosphorylation of glycogen synthesis kinase 3 α/β. Lithium chloride, an activator of the Wnt pathway, also was protective against glucocorticoid-induced apoptosis. Whereas it is known that mechanical loading increases cyclooxygenase-2 and EP2 receptor expression and prostaglandin production, dexamethasone was shown to inhibit expression of these components of the prostaglandin pathway and to reduce β-catenin protein expression. β-catenin siRNA knockdown experiments abrogated the protective effects of PGE(2), confirming the central role of β-catenin in mediating the protection against dexamethasone-induced cell death. Our data support a central role for PGE(2) acting through the cAMP/PKA and β-catenin signaling pathways in the protection of osteocyte apoptosis by fluid-flow shear stress.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)