Merck
  • The cytoplasmic domain of proEGF negatively regulates motility and elastinolytic activity in thyroid carcinoma cells.
The cytoplasmic domain of proEGF negatively regulates motility and elastinolytic activity in thyroid carcinoma cells.
Neoplasia (New York, N.Y.) (2008-09-25)
Aleksandra Glogowska, Janette Pyka, Astrid Kehlen, Marek Los, Paul Perumal, Ekkehard Weber, Sheue-yann Cheng, Cuong Hoang-Vu, Thomas Klonisch
ABSTRACT
The intracellular domains of the membrane-anchoring regions of some precursors of epidermal growth factor (EGF) family members have intrinsic biologic activities. We have determined the role of the human proEGF cytoplasmic domain (proEGFcyt) as part of the proEGF transmembrane-anchored region (proEGFctF) in the regulation of motility and elastinolytic invasion in human thyroid cancer cells. We found proEGFctF to act as a negative regulator of motility and elastin matrix penetration and the presence of proEGFcyt or proEGF22.23 resulted in a similar reduction in motility and elastinolytic migration. This activity was counteracted by EGF-induced activation of EGF receptor signaling. Decreased elastinolytic migratory activity in the presence of proEGFctF and proEGFcyt/proEGF22.23 coincided with decreased secretion of elastinolytic procathepsin L. The presence of proEGFctF and proEGFcyt/proEGF22.23 coincided with the specific transcriptional up-regulation of t-SNARE member SNAP25. Treatment with siRNA-SNAP25 resulted in motility and elastin migration being restored to normal levels. Epidermal growth factor treatment down-regulated SNAP25 protein by activating EGF receptor-mediated proteasomal degradation of SNAP25. These data provide first evidence for an important function of the cytoplasmic domain of the human proEGF transmembrane region as a novel suppressor of motility and cathepsin L-mediated elastinolytic invasion in human thyroid carcinoma cells and suggest important clinical implications for EGF-expressing tumors.
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
Human serum (cystatin C), ERM®, certified reference material