Merck
Dihydromyricetin as a novel anti-alcohol intoxication medication.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-01-06)
Yi Shen, A Kerstin Lindemeyer, Claudia Gonzalez, Xuesi M Shao, Igor Spigelman, Richard W Olsen, Jing Liang
ABSTRACT
Alcohol use disorders (AUDs) constitute the most common form of substance abuse. The development of AUDs involves repeated alcohol use leading to tolerance, alcohol withdrawal syndrome, and physical and psychological dependence, with loss of ability to control excessive drinking. Currently there is no effective therapeutic agent for AUDs without major side effects. Dihydromyricetin (DHM; 1 mg/kg, i.p. injection), a flavonoid component of herbal medicines, counteracted acute alcohol (EtOH) intoxication, and also withdrawal signs in rats including tolerance, increased anxiety, and seizure susceptibility; DHM greatly reduced EtOH consumption in an intermittent voluntary EtOH intake paradigm in rats. GABA(A) receptors (GABA(A)Rs) are major targets of acute and chronic EtOH actions on the brain. At the cellular levels, DHM (1 μM) antagonized both acute EtOH-induced potentiation of GABA(A)Rs and EtOH exposure/withdrawal-induced GABA(A)R plasticity, including alterations in responsiveness of extrasynaptic and postsynaptic GABA(A)Rs to acute EtOH and, most importantly, increases in GABA(A)R α4 subunit expression in hippocampus and cultured neurons. DHM anti-alcohol effects on both behavior and CNS neurons were antagonized by flumazenil (10 mg/kg in vivo; 10 μM in vitro), the benzodiazepine (BZ) antagonist. DHM competitively inhibited BZ-site [(3)H]flunitrazepam binding (IC(50), 4.36 μM), suggesting DHM interaction with EtOH involves the BZ sites on GABA(A)Rs. In summary, we determined DHM anti-alcoholic effects on animal models and determined a major molecular target and cellular mechanism of DHM for counteracting alcohol intoxication and dependence. We demonstrated pharmacological properties of DHM consistent with those expected to underlie successful medical treatment of AUDs; therefore DHM is a therapeutic candidate.
Sigma-Aldrich
Dihydromyricetin, ≥98% (HPLC)