Merck
  • Liquid chromatography-mass spectrometry metabolite library for metabolomics: Evaluating column suitability using a scoring approach.
Liquid chromatography-mass spectrometry metabolite library for metabolomics: Evaluating column suitability using a scoring approach.
Journal of chromatography. A (2023-01-22)
Dimitra Diamantidou, Ioannis Sampsonidis, Theodoros Liapikos, Helen Gika, Georgios Theodoridis
ABSTRACT
Untargeted metabolomic studies require an extensive set of analyte (metabolic) information to be obtained from each analyzed sample. Thus, highly selective, and efficient analytical methodologies together with reversed-phase (RP) or hydrophilic interaction liquid chromatography (HILIC) are usually applied in these approaches. Here, we present a performance comparison of five different chromatographic columns (C18, C8, RP Amide, zicHILIC, OH5 HILIC phases) to evaluate their sufficiency of analysis for a large analyte library, consisting of 817 authentic standards. By taking into account experimental chromatographic parameters (i.e. retention time, peak tailing and asymmetry, FWHM, signal-to-noise ratio and peak area and intensity), the proposed column scoring approach provides a simple criterion that may assist analysis in the select of a stationary phase for those metabolites of interest. RPLC methods offered better results regarding metabolic library coverage, while the zicHILIC stationary phase delivered a bigger number of properly eluted compounds. This study demonstrates the importance of choosing the most suitable configuration for the analysis of different metabolic classes.
Product Description
Supelco
Ascentis® Express RP-Amide, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Sigma-Aldrich
Organic Acid Metabolite Library of Standards, Supplied by IROA Technologies
Sigma-Aldrich
Fatty Acid Metabolite Library of Standards, Supplied by IROA Technologies
Sigma-Aldrich
Mass Spectrometry Metabolite Library, Supplied by IROA Technologies
Sigma-Aldrich
Bile Acid/Carnitine/Sterol Metabolite Library of Standards, Supplied by IROA Technologies
Supelco
SeQuant® ZIC-HILIC 3.5µm,200Å 100 x 2.1 mm, PEEK-lined stainless steel column, suitable for HPLC
Supelco
Ascentis® Express C8, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express OH5, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express 90 Å AQ-C18, 2.7 μm HPLC Column, column L × I.D. 10 cm × 2.1 mm