Merck
  • ERK3/MAPK6 dictates CDC42/RAC1 activity and ARP2/3-dependent actin polymerization.
ERK3/MAPK6 dictates CDC42/RAC1 activity and ARP2/3-dependent actin polymerization.
eLife (2023-04-15)
Katarzyna Bogucka-Janczi, Gregory Harms, Marie-May Coissieux, Mohamed Bentires-Alj, Bernd Thiede, Krishnaraj Rajalingam
ABSTRACT
The actin cytoskeleton is tightly controlled by RhoGTPases, actin binding-proteins and nucleation-promoting factors to perform fundamental cellular functions. We have previously shown that ERK3, an atypical MAPK, controls IL-8 production and chemotaxis (Bogueka et al., 2020). Here, we show in human cells that ERK3 directly acts as a guanine nucleotide exchange factor for CDC42 and phosphorylates the ARP3 subunit of the ARP2/3 complex at S418 to promote filopodia formation and actin polymerization, respectively. Consistently, depletion of ERK3 prevented both basal and EGF-dependent RAC1 and CDC42 activation, maintenance of F-actin content, filopodia formation, and epithelial cell migration. Further, ERK3 protein bound directly to the purified ARP2/3 complex and augmented polymerization of actin in vitro. ERK3 kinase activity was required for the formation of actin-rich protrusions in mammalian cells. These findings unveil a fundamentally unique pathway employed by cells to control actin-dependent cellular functions.
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
Anti-phospho-ERK3 (pSer189) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ARPC1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution