Merck
  • Structure of the endosomal Commander complex linked to Ritscher-Schinzel syndrome.
Structure of the endosomal Commander complex linked to Ritscher-Schinzel syndrome.
Cell (2023-05-13)
Michael D Healy, Kerrie E McNally, Rebeka Butkovič, Molly Chilton, Kohji Kato, Joanna Sacharz, Calum McConville, Edmund R R Moody, Shrestha Shaw, Vicente J Planelles-Herrero, Sathish K N Yadav, Jennifer Ross, Ufuk Borucu, Catherine S Palmer, Kai-En Chen, Tristan I Croll, Ryan J Hall, Nikeisha J Caruana, Rajesh Ghai, Thi H D Nguyen, Kate J Heesom, Shinji Saitoh, Imre Berger, Christiane Schaffitzel, Tom A Williams, David A Stroud, Emmanuel Derivery, Brett M Collins, Peter J Cullen
ABSTRACT
The Commander complex is required for endosomal recycling of diverse transmembrane cargos and is mutated in Ritscher-Schinzel syndrome. It comprises two sub-assemblies: Retriever composed of VPS35L, VPS26C, and VPS29; and the CCC complex which contains twelve subunits: COMMD1-COMMD10 and the coiled-coil domain-containing (CCDC) proteins CCDC22 and CCDC93. Combining X-ray crystallography, electron cryomicroscopy, and in silico predictions, we have assembled a complete structural model of Commander. Retriever is distantly related to the endosomal Retromer complex but has unique features preventing the shared VPS29 subunit from interacting with Retromer-associated factors. The COMMD proteins form a distinctive hetero-decameric ring stabilized by extensive interactions with CCDC22 and CCDC93. These adopt a coiled-coil structure that connects the CCC and Retriever assemblies and recruits a 16th subunit, DENND10, to form the complete Commander complex. The structure allows mapping of disease-causing mutations and reveals the molecular features required for the function of this evolutionarily conserved trafficking machinery.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-COMMD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)
Sigma-Aldrich
Monoclonal Anti-COMMD1 antibody produced in mouse, clone 2A12, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-DSCR3 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)