Merck
  • The Nse5/6-like SIMC1-SLF2 complex localizes SMC5/6 to viral replication centers.
The Nse5/6-like SIMC1-SLF2 complex localizes SMC5/6 to viral replication centers.
eLife (2022-11-15)
Martina Oravcová, Minghua Nie, Nicola Zilio, Shintaro Maeda, Yasaman Jami-Alahmadi, Eros Lazzerini-Denchi, James A Wohlschlegel, Helle D Ulrich, Takanori Otomo, Michael N Boddy
ABSTRACT
The human SMC5/6 complex is a conserved guardian of genome stability and an emerging component of antiviral responses. These disparate functions likely require distinct mechanisms of SMC5/6 regulation. In yeast, Smc5/6 is regulated by its Nse5/6 subunits, but such regulatory subunits for human SMC5/6 are poorly defined. Here, we identify a novel SMC5/6 subunit called SIMC1 that contains SUMO interacting motifs (SIMs) and an Nse5-like domain. We isolated SIMC1 from the proteomic environment of SMC5/6 within polyomavirus large T antigen (LT)-induced subnuclear compartments. SIMC1 uses its SIMs and Nse5-like domain to localize SMC5/6 to polyomavirus replication centers (PyVRCs) at SUMO-rich PML nuclear bodies. SIMC1's Nse5-like domain binds to the putative Nse6 orthologue SLF2 to form an anti-parallel helical dimer resembling the yeast Nse5/6 structure. SIMC1-SLF2 structure-based mutagenesis defines a conserved surface region containing the N-terminus of SIMC1's helical domain that regulates SMC5/6 localization to PyVRCs. Furthermore, SLF1, which recruits SMC5/6 to DNA lesions via its BRCT and ARD motifs, binds SLF2 analogously to SIMC1 and forms a separate Nse5/6-like complex. Thus, two Nse5/6-like complexes with distinct recruitment domains control human SMC5/6 localization.
Sigma-Aldrich
Monoclonal Anti-PSTAIR antibody produced in mouse, clone PSTAIR, ascites fluid
Sigma-Aldrich
Gelatin from cold water fish skin, 40-50% in H2O
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-SUMO-2/3 Antibody, clone 8A2, clone 8A2, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Millipore
Benzonase® Nuclease, Purity > 90%