Merck
  • Targeting MDM4 as a Novel Therapeutic Approach in Prostate Cancer Independent of p53 Status.
Targeting MDM4 as a Novel Therapeutic Approach in Prostate Cancer Independent of p53 Status.
Cancers (2022-08-27)
Javier Octavio Mejía-Hernández, Dinesh Raghu, Franco Caramia, Nicholas Clemons, Kenji Fujihara, Thomas Riseborough, Amina Teunisse, Aart G Jochemsen, Lars Abrahmsén, Giovanni Blandino, Andrea Russo, Cristina Gamell, Stephen B Fox, Catherine Mitchell, Elena A Takano, David Byrne, Panimaya Jeffreena Miranda, Reem Saleh, Heather Thorne, Shahneen Sandhu, Scott G Williams, Simon P Keam, Ygal Haupt, Sue Haupt
ABSTRACT
Metastatic prostate cancer is a lethal disease in patients incapable of responding to therapeutic interventions. Invasive prostate cancer spread is caused by failure of the normal anti-cancer defense systems that are controlled by the tumour suppressor protein, p53. Upon mutation, p53 malfunctions. Therapeutic strategies to directly re-empower the growth-restrictive capacities of p53 in cancers have largely been unsuccessful, frequently because of a failure to discriminate responses in diseased and healthy tissues. Our studies sought alternative prostate cancer drivers, intending to uncover new treatment targets. We discovered the oncogenic potency of MDM4 in prostate cancer cells, both in the presence and absence of p53 and also its mutation. We uncovered that sustained depletion of MDM4 is growth inhibitory in prostate cancer cells, involving either apoptosis or senescence, depending on the cell and genetic context. We identified that the potency of MDM4 targeting could be potentiated in prostate cancers with mutant p53 through the addition of a first-in-class small molecule drug that was selected as a p53 reactivator and has the capacity to elevate oxidative stress in cancer cells to drive their death.
Sigma-Aldrich
Z-VAD-FMK, solid
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Mdmx Antibody, clone 8C6, clone 8C6, from mouse
Sigma-Aldrich
Anti-MDM2 Antibody, clone 3G9, clone 3G9, from mouse
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)