Merck
  • Procedures for derivation and characterisation of human embryonic stem cells from Odense, Denmark.
Procedures for derivation and characterisation of human embryonic stem cells from Odense, Denmark.
Methods in molecular biology (Clifton, N.J.) (2012-04-25)
Linda Harkness, Moustapha Kassem
ABSTRACT
In 1998, a development occurred in stem cell biology with the first report of the derivation of a human embryonic stem cell (hESC) line. Since then a number of techniques have been used to derive and characterise hESCs. Here, we describe the derivation methods used by our laboratory for isolation of the ICM by immunosurgery and outgrowth of the whole blastocyst. We have added protocols for routine culture, passaging and cryopreservation of our hESC lines as well as the methods we have used for characterisation (flow cytometry, karyotyping, immunocytochemistry, in vitro and in vivo differentiation). Additionally, we have included gene sequences for PCR and an antibody list for immunocytochemistry.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Human IgG (whole molecule) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Complement sera from guinea pig, lyophilized powder
Sigma-Aldrich
Anti-TRA-1-85 Antibody, blood group Antigen Ok(a), clone TRA-1-85, clone TRA-1-85, Chemicon®, from mouse