Merck
  • Cardiac fibroblasts regulate the development of heart failure via Htra3-TGF-β-IGFBP7 axis.
Cardiac fibroblasts regulate the development of heart failure via Htra3-TGF-β-IGFBP7 axis.
Nature communications (2022-06-08)
Toshiyuki Ko, Seitaro Nomura, Shintaro Yamada, Kanna Fujita, Takanori Fujita, Masahiro Satoh, Chio Oka, Manami Katoh, Masamichi Ito, Mikako Katagiri, Tatsuro Sassa, Bo Zhang, Satoshi Hatsuse, Takanobu Yamada, Mutsuo Harada, Haruhiro Toko, Eisuke Amiya, Masaru Hatano, Osamu Kinoshita, Kan Nawata, Hiroyuki Abe, Tetsuo Ushiku, Minoru Ono, Masashi Ikeuchi, Hiroyuki Morita, Hiroyuki Aburatani, Issei Komuro
ABSTRACT
Tissue fibrosis and organ dysfunction are hallmarks of age-related diseases including heart failure, but it remains elusive whether there is a common pathway to induce both events. Through single-cell RNA-seq, spatial transcriptomics, and genetic perturbation, we elucidate that high-temperature requirement A serine peptidase 3 (Htra3) is a critical regulator of cardiac fibrosis and heart failure by maintaining the identity of quiescent cardiac fibroblasts through degrading transforming growth factor-β (TGF-β). Pressure overload downregulates expression of Htra3 in cardiac fibroblasts and activated TGF-β signaling, which induces not only cardiac fibrosis but also heart failure through DNA damage accumulation and secretory phenotype induction in failing cardiomyocytes. Overexpression of Htra3 in the heart inhibits TGF-β signaling and ameliorates cardiac dysfunction after pressure overload. Htra3-regulated induction of spatio-temporal cardiac fibrosis and cardiomyocyte secretory phenotype are observed specifically in infarct regions after myocardial infarction. Integrative analyses of single-cardiomyocyte transcriptome and plasma proteome in human reveal that IGFBP7, which is a cytokine downstream of TGF-β and secreted from failing cardiomyocytes, is the most predictable marker of advanced heart failure. These findings highlight the roles of cardiac fibroblasts in regulating cardiomyocyte homeostasis and cardiac fibrosis through the Htra3-TGF-β-IGFBP7 pathway, which would be a therapeutic target for heart failure.
Sigma-Aldrich
Anti-TGF β Receptor II antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)